Learn More
Sigma Organic Chemistry Heptyl 4-hydroxybenzoat | 1G | 1085-12-7 | MFCD00016481
Supplier: Sigma Organic Chemistry CDS0012781G

Heptyl 4-hydroxybenzoat , 1G
About this Item:
CAS #: 1085-12-7
MDL #: MFCD00016481
Molecular Weight: 236.31
UNSPSC Code: 12352200
Chemical Formula: C14H20O3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.