Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptyl 4-Hydroxybenzoate, Spectrum™ Chemical
SDP

Catalog No. 18620839 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-839 25 g
18-620-840 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-620-839 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H153225GM
Add to Cart
Add to Cart

Synonym: Heptylparaben, n-Heptyl 4-Hydroxybenzoate, Heptyl Paraben

Chemical Identifiers

CAS 1085-12-7
Molecular Formula C14H20O3
Molecular Weight (g/mol) 236.31
InChI Key ZTJORNVITHUQJA-UHFFFAOYSA-N
IUPAC Name heptyl 4-hydroxybenzoate
SMILES CCCCCCCOC(=O)C1=CC=C(O)C=C1

Specifications

Quantity 25 g
Percent Purity 98%
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.