Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptylnonyl Adipate (so called) [Plasticizer], TCI America™
SDP

Catalog No. A1388500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1388500ML 500 mL
A138825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1388500ML Supplier TCI America Supplier No. A1388500ML

Chemical Identifiers

MDL Number MFCD00236062
Synonym Adipic Acid Heptylnonyl Ester

Specifications

Color Colorless
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Heptylnonyl Adipate (so called) [Plasticizer]
EINECSNumber (2)-0861
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.