Learn More
Siemens Healthineers Hepzyme™ Reagent
Catalog No. 23044661 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Catalog No. Quantity
23-044-661 10 x 1 mL
1 options
This item is not returnable. View return policy
Catalog No. 23-044-661 Supplier Siemens Healthineers Supplier No. 10445730
Add to Cart
Add to Cart
This item is not returnable. View return policy
Heparin neutralizer in plasma to rule out heparin contamination in coagulation testing.
Hepzyme reagent is used as a heparin neutralizer in plasma to rule out heparin contamination in coagulation testing.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Coagulation Testing
Quantity 10 x 1 mL