Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HER2 Human anti-Human, BUV615, Clone: Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb), BD OptiBuild™
Human Monoclonal Antibody
Supplier: BD Biosciences 756630
Description
HER2 Monoclonal antibody specifically detects HER2 in Human samples. It is validated for Flow cytometry
Specifications
HER2 | |
Monoclonal | |
0.2 mg/mL | |
CD340; ERBB2; HER-2; HER-2/neu; HER2; NEU; proto-oncogene c-ErbB-2 | |
A431 Cell Line Expressing EGFR | |
50 μg | |
Primary | |
Human | |
Liquid |
Flow Cytometry | |
Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human | |
Affinity Chromatography | |
RUO | |
2064 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction