Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HER2 Human anti-Human, BV480, Clone: Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb), BD OptiBuild™

Human Monoclonal Antibody

Supplier:  BD Biosciences 756629

Catalog No. BDB756629


Only null left
Add to Cart

Description

Description

HER2 Monoclonal antibody specifically detects HER2 in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HER2
Monoclonal
0.2 mg/mL
CD340; ERBB2; HER-2; HER-2/neu; HER2; NEU; proto-oncogene c-ErbB-2
A431 Cell Line Expressing EGFR
50 μg
Primary
Human
Liquid
Flow Cytometry
Trastuzumab297.rMAb (also known as Trastuzumab N297A Biosimilar.rMAb)
Aqueous buffered solution containing ≤0.09% sodium azide.
Human
Affinity Chromatography
RUO
2064
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.