Learn More
Diagnostic Biosystems HER2/NEU C-ERBB-2 c-erbB-2 Oncoprotein (SP3)
Supplier:  Diagnostic Biosystems RMAB008
Encompass_Preferred
This antibody recognizes a c-erbB-2 protein, which is a receptor tyrosine kinase of the c-erbB family. It is closely related in structure to the epidermal growth factor receptor. c-erbB-2 oncoprotein is detectable in a subset of breast and other adenocarcinomas, as well astransitional cell carcinomas. In the case of breast cancer, expression determined byimmunohistochemistry has been shown to be associated with poor prognosis.
Catalog No. NC2115335
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.