Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Herasafe™ 2030i CONFIGURED-NSF Biosafety Cabinet PROMO

Catalog No. 09034250PM
Change view
Click to view available options
Dimensions (L x W x H) Exterior:
80 x 130 x 154 cm
Outlets:
2 Rear Wall
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Exterior Outlets
09-034-250PM 80 x 130 x 154 cm 2 Rear Wall
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Attention:This item requires special delivery information. You will be asked for this during the checkout process.
Catalog No. 09-034-250PM Supplier Thermo Scientific™ Supplier No. 51032334TU
Attention:This item requires special delivery information. You will be asked for this during the checkout process.

Herasafe 2030i incorporates SmartFlow Plus, DAVe Plus and Smart UV technologies for ultimate protection and graphical user interface for ease of operation. Unit includes inbuilt Wi-Fi capability and is ready to be 'connected'.

Breakthrough innovations and unprecedented features make the Herasafe 2030i BSC Thermo Scientific™'s most technologically advanced and user-friendly Class II, Type A2 cabinet available. (Stand sold separately)

Smart protection:

  • Thermo Scientific™ SmartFlow™ Plus Technology - Autonomous safety features maintain proper air flow and monitor critical conditions in real time without user interaction.
  • Self-adjusting germicidal UV light intensity - Unique cross beam UV system has exclusive self-adjusting exposure capability to enable reproducible germicidal effectiveness with every use
  • 316L stainless steel interior - robust and resistant to scratches and chemicals

Simplified operation and ergonomics:

  • Touchscreen interface
  • SmartClean Plus window design – Easily access the interior of the BSC with the motorized front sash which automatically stops at the set working height. The sash can be further opened electronically or the whole front panel hinges open for thorough cleaning access.
  • Bright work area with glass side walls and LED lights
  • Ergonomic Arm Rests (option)
  • SmartPort convenience - Easily pass tubing and wiring through sidewall to accommodate many applications needs.

Information management:

  • Touchscreen and connectivity (available in select countries) – Built-in WiFi system is ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools and more. Digital Airflow Verification (DAVe) alerts, system self-checks and performance status are provided on the full-color touchscreen.

Energy efficiency:

  • LED interior illumination
  • Dual DC motors
  • Standby mode

Certifications:

  • For North America, cabinets are NSF-49 certified (4 ft. and 6 ft. models)
  • For Europe, the cabinets are certified according to EN12469 (all 4 sizes). The 1.2 m model has been certified to EN 12469 with the NF mark by LNE as well.

Available sizes:

  • North America 4 ft. and 6 ft. models; other countries: 0.9 m, 1.2 m, 1.5 m and 1.8 m

For cell and gene therapy applications, we offer the Herasafe 2030i Biological Safety Cabinet CTS (Cell Therapy Systems) Series which consists of the product, a comprehensive documentation package including certifications and protocols, as well as compliance services.

Previous models: Herasafe KS Note: Please order stand separately and indicate any additional requirements/ options you need when you 'Request a quote.'

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NSF/ANSI 49
Chamber Material Stainless Steel (316)
Product Type Biosafety Cabinet
Model Herasafe 2030i
Outlets 2 Rear Wall
Sash Opening (Metric) 25.4 cm
Display 7 in. GUI
Filter Efficiency 0.99995
Filter Type H14 HEPA
Green Features Energy Efficient
Energy Consumption (Reduced Flow Mode) 55 W/hr.
Lighting LED
Ports 2 Ports/Side with Additional Smart Ports
Sash Opening (English) 10 in.
Dimensions (L x W x H) Exterior 80 x 130 x 154 cm
Frequency 60 Hz
Net Weight (English) 441 lb.
Weight (English) 496 lb.
Electrical Requirements 120 V
Energy Consumption (Operating Set Point) 180 W/hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.