Learn More
Thermo Scientific™ Heratherm™ General Protocol Ovens

Perform routine, day-to-day work in these general protocol ovens, which provide a safe, ideal heating and drying solution for research, clinical or industrial needs.
Supplier: Thermo Scientific™ 51028875
Description
Perform routine applications simply and safely with Thermo Scientific™ Heratherm™ General Protocol Ovens. These ovens provide an ideal heating and drying solution for research, clinical or industrial needs.
Features:
- Operating temperatures from 50°C to 250°C
- Automatic over-temperature alarm system
- Low overall energy consumption
- Built-in timer
- Corrosion-resistant stainless steel chambers with rounded corners
- Large, easy-to-read vacuum fluorescent display
- Easy-to-use, microprocessor-controlled touch button operation
- Doors can be opened at over 180°C
Ordering Information:
Availability may vary by country

Specifications
Microprocessor control with vacuum fluorescent display | |
50°C to 250°C | |
2 supplied/19 max. | |
ON / OFF | |
73.8 x 64 x 92 cm | |
73.8 cm | |
18.3 in. | |
46.4 cm | |
27.9 in. | |
70.8 cm | |
54.3 cm | |
120 V | |
14 A |
179 L | |
Mechanical Convection Oven | |
±0.3°C | |
21.4 x 18.3 x 27.9 in. (54.3 x 46.4 x 70.8 cm) | |
29.1 in. | |
25.2 in. | |
64 cm | |
36.2 in. | |
92 cm | |
21.4 in. | |
120 V 60 Hz | |
3360 w | |
60 Hz |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.