Learn More
Enzo Life Sciences Herbimycin A (1mg). CAS: 70563-58-5
SDP
Supplier:  Enzo Life Sciences BMLEI2271000
Encompass_Preferred
Antibiotic. Cell permeable tyrosine kinase inhibitor. It is effective on Src, Yes, Fps, Ros, Abl and ErbB oncogene products and inhibits thrombin-induced tyrosine phosphorylation of phospholipase C. It also inhibits HSP90, NF-κB activation and angiogenesis. Increases the sensitivity of certain cancer cells to chemotherapeutic agents. Blocks mitotic activity. Neurotoxic. Formulation: Lyophilized. Purity: ≥98% (HPLC). Solubility: Soluble in ethanol, methanol, DMF or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-0619
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.