Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HERC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562033 Supplier Invitrogen™ Supplier No. PA562033
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KGKLLSSQEG MISFDCFMTI LMQMRRSLGS SADRSQWREP TRTSDGLCSL YEAALCLFEE VCRMASDYSR TCASPDSIQT GDAPIVSETC EVYVWGSNSS HQLVEGTQEK ILQPK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

This gen encodes a member of the HERC protein family. This protein stimulates guanine nucleotide exchange on ARF1 and Rab proteins. This protein may be involved in membrane transport processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HERC1
Gene Accession No. Q15751
Gene Alias guanine nucleotide exchange factor p532; hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1 (CHC1)-like domain (RLD) 1; HECT and RLD domain containing E3 ubiquitin protein ligase family member 1; HECT domain and RCC1-like domain-containing protein 1; HECT-type E3 ubiquitin transferase HERC1; HERC1; p532; p619; Probable E3 ubiquitin-protein ligase HERC1
Gene Symbols HERC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HERC1. Recombinant protein control fragment (Product #RP-98030).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8925
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.