Learn More
Research Products International Corp HERMLE CENTRIFUGE Z207-A-CMB
SDP
Supplier:  Research Products International Corp 260160
Encompass_Preferred
The Z207-A Clinical Centrifuge includes a 8 x 15 ml rotor, and is the perfect centrifuge for any clinical laboratory. The Z207-A also features the unique EZ-Scroll touch pad which gives you ease of control and a modern feel. An advanced microprocessor control allow for setting/displaying of the rotational speed in either rpm or rcf in increments of 10. In addition to retrieving saved programs, the easy to use front panel can be used to adjust accel/decel rates, run time, and change the end of run alarm tone. There is also a momentary spin function that will run while the button is pressed. Additionally the Z207-A features a sleep mode to save energy by shutting off after a certain time if not being used while on, which can be toggled on or off easily.
Catalog No. NC2282240
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.