Learn More
Bioworld Herpes Simplex Virus-2 gD (Recombinant) , 100 μg
Supplier:  Bioworld 220601861
Encompass_Preferred
Recombinant Herpes Simplex Virus-2 glycoprotein D (HSV-2-gD) Source: E.coli Description : Recombinant HSV-2-gD contains the immunodominant regions of glycoprotein D, amino acids: (266-394). HSV-1-gD is purified by proprietary chromatographic techniques. Formulation: 1mg/ml in 25mM Tris-HCl, 1mM EDTA, 50% glycerol. Purity: Greater than 95.0% as determined by: (a) 10% PAGE coomassie staining. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -20C. Please avoid freeze-thaw cycles. Stability: Five years frozen. One month in solution at room temperature Specificity: Immunoreactive with sera of HSV-infected individuals. Applications: 1. ELISA 2. Western blots 3. HSV Detection. Usage: BioWORLD\\\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-286
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.