Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Herpes Simplex Virus (HSV) Type 1 Culture Fluid
SDP

Catalog No. 22157719 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Strain Designations:
Isolate 1
Isolate 10
Isolate 11
Isolate 12
Isolate 13
Isolate 14
Isolate 15
Isolate 16
Isolate 17
Isolate 18
Isolate 19
Isolate 2
Isolate 20
Isolate 21
Isolate 3
Isolate 4
Isolate 5
Isolate 6
Isolate 7
Isolate 8
Isolate 9
Strain Macintyre
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Strain Designations
22-157-719 Isolate 2
22-157-549 Isolate 1
22-157-548 Strain Macintyre
22-157-720 Isolate 3
22-157-721 Isolate 4
22-157-723 Isolate 6
22-157-722 Isolate 5
22-157-724 Isolate 7
22-157-725 Isolate 8
22-157-727 Isolate 10
22-157-726 Isolate 9
22-157-728 Isolate 11
22-157-730 Isolate 13
22-157-731 Isolate 14
22-157-729 Isolate 12
22-157-733 Isolate 16
22-157-732 Isolate 15
22-157-734 Isolate 17
22-157-737 Isolate 20
22-157-735 Isolate 18
22-157-736 Isolate 19
22-157-738 Isolate 21
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Contains live and infectious organisms.
Catalog No. 22-157-719 Supplier ZeptoMetrix Supplier No. 0810183CF
Add to Cart
Add to Cart
Contains live and infectious organisms.

Infectious virus preparation designed to meet research and product development needs

Human Herpes viruses are enveloped icosahedral viruses 150 to 200nm in diameter and contain a nucleocapsid and double-stranded linear DNA.

  • Consist of virus, cells and media taken directly from tissue culture flask
  • Propagated in Vero cell line
  • Biosafety Level 2 organism
  • Available in 1.0mL aliquots
  • Each lot assayed for its Tissue Culture Infective Dose (TCID50)
  • Titer:>105 U/mL

IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.

Nucleic acid/molecular testing, limit of detection (LOD) studies, cross-reactivity studies and other viral-based assays

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Concentration >105U/mL
Content And Storage -65°C
Form Solution
Media Type Vial
Product Type Herpes Simplex Virus Infectious Culture Fluid
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity 1 mL
Strain Designations Isolate 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.