Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes simplex virus Rabbit anti-Herpes simplex virus, Polyclonal, Abnova™

Catalog No. 89033705 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-705 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-705 Supplier Abnova Corporation Supplier No. PAB29860
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against herpes simplex virus (HSV).

  • High specificity
  • High reactivity

Specifications

Antigen Herpes simplex virus
Applications ELISA, Immunofluorescence, Immunohistochemistry, Lateral Flow, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against herpes simplex virus (HSV).
Dilution ELISA Immunofluorescence Immunohistochemistry Lateral-Flow Immunochromatographic Assay Western Blot The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS, pH 7.2 (0.09% sodium azide).
Host Species Rabbit
Immunogen Herpes simplex virus strain F.
Purification Method IgG purification
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with ICPs and late structural proteins of herpes simplex virus.
Target Species Herpes Simplex virus (HSV)
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.