Learn More
ZeptoMetrix Herpes Simplex Virus Type 2 Strain Culture Fluid
Shop All ZeptoMetrix Corporation Products

Description
Human Herpes viruses are enveloped icosahedral viruses 150 to 200nm in diameter that contain a nucleocapsid and double-stranded linear DNA.
- Consist of virus, cells and media taken directly from tissue culture flask
- Propagated in Vero cell line
- Biosafety Level 2 organism
- Available in 1.0mL aliquots
- Each lot assayed for its Tissue Culture Infective Dose (TCID50)
- Titer:>105U/mL
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Nucleic acid/molecular testing, Limit of Detection (LOD) studies, Cross-reactivity studies, Other viral-based assays
Specifications
Specifications
| Biosafety Level | 2 |
| Certifications/Compliance | ISO, 13485 |
| Concentration | >105U/mL |
| Content And Storage | -65°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | Herpes Simplex Virus Infectious Culture Fluid |
| For Use With (Application) | Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
| Quantity | 1 mL |
| Strain Designations | Isolate 10 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.