Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HERVK-113 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144590
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144590 Supplier Invitrogen™ Supplier No. PA5144590
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HERVK-113 Polyclonal Antibody for Western Blot, ELISA

The products of the Gag polyproteins of infectious retroviruses perform highly complex orchestrated tasks during the assembly, budding, maturation, and infection stages of the viral replication cycle. During viral assembly, the proteins form membrane associations and self-associations that ultimately result in budding of an immature virion from the infected cell. Gag precursors also function during viral assembly to selectively bind and package two plus strands of genomic RNA. Endogenous Gag proteins may have kept, lost or modified their original function during evolution.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HERVK-113
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene HERVK_113
Gene Accession No. P63132
Gene Alias Endogenous retrovirus group K member 113 Gag polyprotein; HERVK_113
Gene Symbols HERVK_113
Host Species Rabbit
Immunogen Recombinant Human HERVK_113 protein (283-532aa).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 0
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.