Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Hesperadin hydrochloride
GREENER_CHOICE

Catalog No. 398810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-881-0 10 mg
39-885-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 39-881-0 Supplier Tocris Bioscience™ Supplier No. 3988/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent Aurora kinase B inhibitor

ATP-competitive inhibitor of Aurora B kinase (IC50 = 250 nM). Prevents chromosome alignment and segregation; also induces polyploidy and prevents histone H3-Ser10 phosphorylation. Overrides the spindle assembly checkpoint and induces mitotic exit in monastrol - and taxol -treated HeLa cells.

Chemical Identifiers

CAS 422513-13-1
Molecular Formula C29H32N4O3S

Specifications

Quantity 10 mg
Formula Weight 553.12
Purity >99%
Molecular Weight (g/mol) 516.66
Chemical Name or Material Hesperadin hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.