Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hesperetin 97.0+%, TCI America™
SDP

Catalog No. H072125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H072125G 25 g
H07215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H072125G Supplier TCI America Supplier No. H072125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 520-33-2
Molecular Formula C16H14O6
Molecular Weight (g/mol) 302.28
MDL Number MFCD00075646
InChI Key AIONOLUJZLIMTK-AWEZNQCLSA-N
Synonym hesperetin, hesperitin, 3',5,7-trihydroxy-4'-methoxyflavanone, hesperin, yso2, cyanidanon 4'-methyl ether 1626, 5,7,3'-trihydroxy-4'-methoxyflavanone, --hesperetin, prestwick_908, unii-q9q3d557f1
PubChem CID 72281
ChEBI CHEBI:28230
IUPAC Name (2S)-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
SMILES COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point 232°C
Color White-Yellow
Quantity 25 g
Formula Weight 302.28
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hesperetin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.