Learn More
Sigma Organic Chemistry Hexa(ethylene glycol) dithiol | 500MG | 89141-22-0 | MFCD18632582 | >97%
Supplier: Sigma Organic Chemistry 734616500MG

Hexa(ethylene glycol) dithiol, 500MG
About This Item:
Density: 1.113 g/mL at 25 C
Refractive Index: n20/D 1.495
Storage: 2-8C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.