Learn More
CPC Scientific H-His-His-His-His-His-His-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific MISC014A
Encompass
SEQUENCE: H-His-His-His-His-His-His-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: HHHHHHMOLECULAR FORMULA: C36H44N18O7MOLECULAR WEIGHT: 840.85STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [64134-30-1]SYNONYMS: His Tag, Histidine hexapeptide, SCHEMBL14604365, CTK2A7197, DTXSID10435849, HY-P0294RESEARCH AREA: MiscellaneousREFERENCES:Wieneke, R. et al. JACS. 136, 13975-78 (2014)Liou J-S. et al. Peptides. 37, 273-284 (2012)SKU(s): MISC-014A
Catalog No. 50-210-9740
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.