Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexachlorocyclopentadiene, EPA Individual Analyte, ISO 17034, Methanol, 1 mL

Catalog No. USEPA1127
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-EPA-1127 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-EPA-1127 Supplier Agilent Technologies Supplier No. EPA11271
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 77-47-4 , 67-56-1
Molecular Formula C5Cl6
Molecular Weight (g/mol) 272.755
InChI Key VUNCWTMEJYMOOR-UHFFFAOYSA-N
PubChem CID 6478
IUPAC Name 1,2,3,4,5,5-hexachlorocyclopenta-1,3-diene
SMILES C1(=C(C(C(=C1Cl)Cl)(Cl)Cl)Cl)Cl

Specifications

CAS Max % 99.36, 0.63
Analyte or Component Names methanol,hexachlorobutadiene
Element Methanol, Hexachlorobutadiene
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 5000μg/mL
Chemical Name or Material Hexachlorocyclopentadiene Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.