Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexachlorodisilane 98.0+%, TCI America™
SDP

Catalog No. H102825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H102825G 25 g
H10285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H102825G Supplier TCI America Supplier No. H102825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13465-77-5
Molecular Formula Cl6Si2
Molecular Weight (g/mol) 268.87
MDL Number MFCD00011599
InChI Key LXEXBJXDGVGRAR-UHFFFAOYSA-N
PubChem CID 83497
IUPAC Name trichloro(trichlorosilyl)silane
SMILES [Si]([Si](Cl)(Cl)Cl)(Cl)(Cl)Cl

Specifications

Color Colorless
Quantity 25 g
UN Number 2987
Formula Weight 268.87
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Hexachlorodisilane
EINECSNumber (1)-1214
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.