Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexachloroethane 99.0+%, TCI America™
SDP

Catalog No. H0060100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0060100G 100 g
H006025G 25 g
H0060500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0060100G Supplier TCI America Supplier No. H0060100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-72-1
Molecular Formula C2Cl6
Molecular Weight (g/mol) 236.72
MDL Number MFCD00000799
InChI Key VHHHONWQHHHLTI-UHFFFAOYSA-N
Synonym hexachloroethane, perchloroethane, ethane, hexachloro, avlothane, carbon hexachloride, hexachlorethane, mottenhexe, distokal, distopan, distopin
PubChem CID 6214
ChEBI CHEBI:39227
IUPAC Name hexachloroethane
SMILES ClC(Cl)(Cl)C(Cl)(Cl)Cl

Specifications

Melting Point 185°C
Color White
Quantity 100 g
UN Number 3077
Formula Weight 236.72
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexachloroethane
EINECSNumber (2)-0057
RTECSNumber KI4025000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.