Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexacosane 99.0+%, TCI America™
SDP

Catalog No. H005025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H005025G 25 g
H00505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H005025G Supplier TCI America Supplier No. H005025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 630-01-3
Molecular Formula C26H54
Molecular Weight (g/mol) 366.72
MDL Number MFCD00009354
InChI Key HMSWAIKSFDFLKN-UHFFFAOYSA-N
Synonym n-hexacosane, pentacosane, methyl, ch3-ch2 24-ch3, hexacosane, analytical standard, hexacosane, acmc-1b86y
PubChem CID 12407
ChEBI CHEBI:32940
IUPAC Name hexacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 57°C
Color White
Boiling Point 268°C
Quantity 25 g
Formula Weight 366.72
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexacosane
EINECSNumber (2)-0010
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.