Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecane, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014559 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4559 25 mL
11-101-4560 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-4559 Supplier MilliporeSigma Supelco Supplier No. 5220925ML
Add to Cart
Add to Cart

Hexadecane can find applications as a sole carbon source for the growth of canals, which are supramolecular structures observed in the cell wall of Candida maltose.

Featured Industries: Petroleum
TRUSTED_SUSTAINABILITY

Specifications

CAS 544-76-3
Ignition Point 395°F
Vapor Pressure 1 mmHg (105.3°C)
Vapor Density 7.8 (vs. air)
Density 0.773 g/mL (at 25°C)
Flash Point 135°C
Physical Form Neat
Linear Formula CH3(CH2)14 CH3
MDL Number MFCD00008998
Melting Point 18°C
Boiling Point 287°C
Synonym n-Hexadecane; Cetane
Type Analytical Standard
Quantity 25 mL
Formula Weight 226.44
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.