Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecanedioic Acid 97.0+%, TCI America™
SDP

Catalog No. H029325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H029325G 25 g
H02931G 1 g
H02935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H029325G Supplier TCI America Supplier No. H029325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 505-54-4
Molecular Formula C16H30O4
Molecular Weight (g/mol) 286.412
MDL Number MFCD00002746
InChI Key QQHJDPROMQRDLA-UHFFFAOYSA-N
Synonym thapsic acid, 1,14-tetradecanedicarboxylic acid, 1,16-hexadecanedioic acid, hexadecane-1,16-dioic acid, 1,14-tetradecane dicarboxylic acid, n-tetradecane-.omega.,.omega.'-dicarboxylic acid, dicarboxylic acid c16, 1,16-hexadecanedioate, thapsate, thaspic acid
PubChem CID 10459
ChEBI CHEBI:73722
IUPAC Name hexadecanedioic acid
SMILES C(CCCCCCCC(=O)O)CCCCCCC(=O)O

Specifications

Melting Point 126°C
Color White
Quantity 25 g
Formula Weight 286.41
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecanedioic Acid
EINECSNumber (2)-3082
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.