Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecanophenone 95.0+%, TCI America™
SDP

Catalog No. H05905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05905G 5 g
H059025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05905G Supplier TCI America Supplier No. H05905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6697-12-7
Molecular Formula C22H36O
Molecular Weight (g/mol) 316.53
MDL Number MFCD00008997
InChI Key IIOLAWJMOGLOIB-UHFFFAOYSA-N
Synonym Palmitophenone, Pentadecyl Phenyl Ketone
PubChem CID 81194
IUPAC Name 1-phenylhexadecan-1-one
SMILES CCCCCCCCCCCCCCCC(=O)C1=CC=CC=C1

Specifications

Melting Point 59°C
Color White-Yellow
Boiling Point 248°C
Quantity 5 g
Formula Weight 316.53
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecanophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.