Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyl Acrylate (stabilized with MEHQ) 90.0+%, TCI America™
SDP

Catalog No. H1168250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1168250G 250 g
H116825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1168250G Supplier TCI America Supplier No. H1168250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13402-02-3
Molecular Formula C19H36O2
Molecular Weight (g/mol) 296.50
MDL Number MFCD00078441
InChI Key PZDUWXKXFAIFOR-UHFFFAOYSA-N
Synonym Acrylic Acid Hexadecyl Ester
PubChem CID 83410
IUPAC Name hexadecyl prop-2-enoate
SMILES CCCCCCCCCCCCCCCCOC(=O)C=C

Specifications

Color Yellow
Quantity 250 g
Formula Weight 296.50
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Hexadecyl Acrylate (stabilized with MEHQ)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.