Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadecyl Ether 95.0+%, TCI America™
SDP

Catalog No. H007525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H007525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H007525G Supplier TCI America Supplier No. H007525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4113-12-6
Molecular Formula C32H66O
Molecular Weight (g/mol) 466.88
MDL Number MFCD00026650
InChI Key FDCJDKXCCYFOCV-UHFFFAOYSA-N
Synonym Dihexadecyl Ether
PubChem CID 77757
IUPAC Name 1-hexadecoxyhexadecane
SMILES CCCCCCCCCCCCCCCCOCCCCCCCCCCCCCCCC

Specifications

Melting Point 55°C
Color White-Yellow
Quantity 25 g
Formula Weight 466.88
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Hexadecyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.