Learn More
Sigma Organic Chemistry hexadecyl lactate | 500mg | 35274-05-6 | MFCD00152336
Supplier: Sigma Organic Chemistry CDS003289500MG

hexadecyl lactate , 500mg
About this Item:
CAS #: 35274-05-6
MDL #: MFCD00152336
Molecular Weight: 314.5
UNSPSC Code: 12352200
Chemical Formula: C19H38O3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.