Learn More
Cayman Chemical TrItonX100StockSolutIon250mL
SDP
Supplier:  Cayman Chemical 600217250 ML
Encompass_Preferred
A DMNT1 inhibitor (IC50 0.23 UM) selective for DMNT1 over DMNT3A/3L and DMNT3B/3L and a panel of 34 methyltransferases and 369 kinases (IC50s >10 UM for all) inhibits the proliferation of 51 hematological cancer cell lines (median IC50 0.64 UM) decreases overall DNA methylation and gene expression in MV4-11 THP-1 and NOMO-1 cells in a time- and concentration-dependent manner increases the expression of genes encoding CXCL11 IFI27 and MAGEA4 reduces tumor volume and increases survival but does decrease blood numbers of neutrophils and red blood cells in SKM-1 and MV4-11 leukemia mouse xenograft models at 45 30 or 15 mg/kg twice per day
Catalog No. 50-289-5574
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.