Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexadimethrine Bromide, Spectrum™ Chemical
SDP

Catalog No. 18620893 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-893 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-620-893 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. H341150GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: Polybrene, 6,3-Ionene Bromide, 1,3-Dibromopropane, N,N,N',N'-Tetramethylhexane-1,6-Diamine

Hexadimethrine Bromide, also known as Polybrene, is a cationic polymer that plays an important a role in protein sequencing. Non-graded Hexadimethrine Bromide supplied by Spectrum™ is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Chemical Identifiers

CAS 28728-55-4

Specifications

Packaging Bottle
Quantity 50 g
Percent Purity 90%
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.