accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3NCH3CH3BrBr

CAS RN 28728-55-4

IUPAC Name: 1,3-dibromopropane; [6-(dimethylamino)hexyl]dimethylamine
Synonyms: polybrene
Molecular Weight (g/mol): 374.21
Molecular Formula: C13H30Br2N2
InChi Key: KZKAYEGOIJEWQB-UHFFFAOYSA-N
SMILES: BrCCCBr.CN(C)CCCCCCN(C)C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)