Learn More
Hexaethylene glycol, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Hexaethylene glycol exhibits antioxidant, antimicrobial and anti-cancer properties. It also shows potential application as functional hydraulic fluids. It is used as a surfactant building block as well as used in biological sample preparation. Further, it is employed in the synthesis of binaphthol-based macrocyclic ethers using intramolecular oxidative coupling with CuCl(OH).
Solubility
Miscible with chloroform and methanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 5°C to 7°C |
| Density | 1.123 |
| Boiling Point | 216°C to 217°C (4 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.464 |
| Quantity | 5 g |
| Beilstein | 1638281 |
| Solubility Information | Miscible with chloroform and methanol. |
| Formula Weight | 282.33 |
| Percent Purity | 96% |
| Show More |
Safety and Handling
| EINECSNumber | 220-045-1 |
| RTECSNumber | MM3670000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.