Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexafluoroacetone Hydrate 97.0+%, TCI America™
SDP

Catalog No. H042525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H042525G 25 g
H04255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H042525G Supplier TCI America Supplier No. H042525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34202-69-2
Molecular Formula C3H6F6O4
Molecular Weight (g/mol) 220.067
MDL Number MFCD00149061
InChI Key SNZAEUWCEHDROX-UHFFFAOYSA-N
Synonym hexafluoroacetone trihydrate, 1,1,1,3,3,3-hexafluoropropan-2-one trihydrate, 2-propanone, hexafluoro-, trihydrate, hfa trihydrate, acetone, hexafluoro-, trihydrate, 2-propanone, 1,1,1,3,3,3-hexafluoro-, trihydrate, perfluoroacetone trihydrate, hexafluoro-2-propanone trihydrate, hexafluoro-2-propanone trihydrate; perfluoroacetone trihydrate, hexafluoroacetonetrihydrate
PubChem CID 36719
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-one;trihydrate
SMILES C(=O)(C(F)(F)F)C(F)(F)F.O.O.O

Specifications

Color Yellow
Quantity 25 g
UN Number 2552
Formula Weight 166.02
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Hexafluoroacetone Hydrate
EINECSNumber (2)-0581
RTECSNumber UC2700000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.