Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexafluorobenzene, 99%

Catalog No. p-3778741
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120540100 10 mL Glass bottle
AC120541000 100 mL Glass bottle
AC120545000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC120540100 Supplier Thermo Scientific Chemicals Supplier No. 120540100
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 392-56-3 | C6F6 | 186.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-56-3
Molecular Formula C6F6
Molecular Weight (g/mol) 186.06
MDL Number MFCD00000288
InChI Key ZQBFAOFFOQMSGJ-UHFFFAOYSA-N
Synonym hexafluorobenzene, perfluorobenzene, benzene, hexafluoro, hexafluorbenzol, unii-cmc18t611k, benzene, 1,2,3,4,5,6-hexafluoro, hexa fluorobenzene, hexafluoro benzene, pubchem18879, acmc-1bmus
PubChem CID 9805
ChEBI CHEBI:38589
IUPAC Name 1,2,3,4,5,6-hexafluorobenzene
SMILES C1(=C(C(=C(C(=C1F)F)F)F)F)F

Specifications

Melting Point 3.7°C to 4.1°C
Color Colorless to Yellow
Density 1.6120g/mL
Boiling Point 81.0°C to 82.0°C (743.0 mmHg)
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.3760 to 1.3780
Linear Formula C6F6
Quantity 10 mL
Merck Index 15,4722
Specific Gravity 1.612
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in ethanol
Formula Weight 186.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexafluorobenzene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
EINECSNumber 206-876-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.