Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexafluorocyclotriphosphazene 97.0+%, TCI America™
SDP

Catalog No. H142925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H142925G 25 g
H14295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H142925G Supplier TCI America Supplier No. H142925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15599-91-4
Molecular Formula F6N3P3
Molecular Weight (g/mol) 248.93
MDL Number MFCD00192400
InChI Key DKQPXAWBVGCNHG-UHFFFAOYSA-N
PubChem CID 139958
IUPAC Name hexafluoro-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
SMILES FP1(F)=NP(F)(F)=NP(F)(F)=N1

Specifications

Melting Point 27°C
Color White-Yellow
Boiling Point 51°C
Quantity 25 g
UN Number 1759
Formula Weight 248.93
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Hexafluorocyclotriphosphazene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.