Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Hexagonal Base Graduated Cylinder
SDPSCI_ED

Catalog No. S01343 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 ml
250 mL
500 mL
1000 mL
2000 mL
Graduations:
0.2 mL
0.5 mL
1 mL
2 mL
5 mL
10 mL
20 mL
Height (Metric):
110 mm
170 mm
198 mm
210 mm
321 mm
361 mm
375 mm
535 mm
Quantity:
1 Ea.
12 Pk.
2 Pk.
24 Pk.
3 Pk.
4 Pk.
6 Pk.
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Graduations Height (Metric) Quantity
S01343 10 mL 0.2 mL 110 mm 1 Ea.
S01157 10 mL 0.2 mL 110 mm 24 Pk.
S01342 25 mL 0.5 mL 170 mm 1 Ea.
S01158 25 mL 0.5 mL 170 mm 12 Pk.
S01341 50 mL 1 mL 198 mm 1 Ea.
S01159 50 mL 1 mL 198 mm 12 Pk.
S01340 100 ml 1 mL 210 mm 1 Ea.
S01160 100 ml 1 mL 210 mm 12 Pk.
S01339 250 mL 2 mL 321 mm 1 Ea.
S01161 250 mL 2 mL 321 mm 6 Pk.
S01338 500 mL 5 mL 361 mm 1 Ea.
S01162 500 mL 5 mL 361 mm 4 Pk.
S01337 100 ml 10 mL 375 mm 1 Ea.
S01163 1000 mL 10 mL 375 mm 3 Pk.
S27766 2000 mL 20 mL 535 mm 2 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. S01343 Supplier United Scientific™ Supplier No. 80031EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Measuring cylinder with raised graduations, contact clarity, and hexagonal base for stability.

  • Polypropylene one-piece measuring cylinders have raised graduations for easy readability
  • They are autoclavable and have excellent contact clarity
  • The hexagonal base provides excellent stability to the cylinder, and the tapered spout enables easy pouring of reagents

Specifications

Capacity (Metric) 10 mL
Material Polypropylene
Diameter (English) 0.6 in.
Diameter (Metric) 15 mm
Quantity 1 Ea.
Graduations 0.2 mL
Base Type Hexagonal
Height (English) 4.3 in.
Height (Metric) 110 mm
Product Type Graduated Cylinder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.