Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hexamethylbenzene-d18 | 4342-40-9 | MFCD00144445 | 99.5% | 180.38 g·mol-1 | C12D18 | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW038500S10MG

Encompass_Preferred

Hexamethylbenzene-d18 is the fully deuterated analog of hexamethylbenzene used as an isotopic tracer and reference standard for analytical and research applications. Supplied as a purified powder, it is intended for laboratory use where high isotopic enrichment and chemical purity are required.

  • High isotopic enrichment (99.3% D).
  • High chemical purity (99.5% by HPLC).
  • Stable powder form for convenient storage and handling.
  • Suitable as an internal standard or tracer in mass spectrometry and quantitative studies.
  • Recommended storage conditions extend shelf life.

Catalog No. 50-003-01205


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.