Learn More
Medchemexpress LLC Benzene, 1,2,3,4,5,6-hexa(methyl-d3)- | 4342-40-9 | MFCD00144445 | 99.5% | 180.38 | C12D18 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW038500S50MG
Encompass_Preferred
Hexamethylbenzene-d18 is the fully deuterated isotopologue of hexamethylbenzene (C12D18, MW 180.38, CAS 4342-40-9) used as a stable isotope reference for spectroscopic, mechanistic, and tracer studies. It is provided in small research-scale quantities and accompanied by product documentation for analytical use.
  • Fully deuterated isotopologue for isotope labeling studies.
  • High purity appropriate for spectroscopic and analytical applications.
  • Provided with certificate of analysis for batch-specific characterization.
  • Available in small milligram sizes for research workflows.
  • Suitable as an internal standard for NMR and mass spectrometry.
Catalog No. 50-003-00373
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.