Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethyldisilazane, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018339 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8339 250 mL
11-101-9816 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8339 Supplier MilliporeSigma Supelco Supplier No. 52619250ML
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00008259
Synonym HMDS

Specifications

Boiling Point 125°C
CAS 999-97-3
Quantity 250 mL
Linear Formula (CH3)3 SiNHSi(CH3)3
Flash Point 11°C
Formula Weight 161.39
Percent Purity ≥99.0% (GC)
Grade GC Derivatization
RTECSNumber JM9230000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.