Learn More
Sigma Aldrich Fine Chemicals Biosciences Hexamethyldisilazane produ5KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 86944
Encompass
Hexamethyldisilazane (HDMS) is a bulk organo silicon compound.It finds major application as a silanizing agent silylation with HMDS is carried out in the presence of an acid catalyst.The hydrophobic nature of HDMS coatings on various nanoparticles renders them resistant to water contamination and flocculation during synthesis.
Catalog No. 50-290-6732
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.