Learn More
Sigma Organic Chemistry Hexamethylenediamine | 25G | 124-09-4 | MFCD00008243
Supplier: Sigma Organic Chemistry H1169625G

Hexamethylenediamine, 25G
About This Item:
Synonym(s): 1,6-Diaminohexane; 1,6-Hexanediamine; HMDA
Linear Formula: NH2(CH2)6NH2
Molecular Weight: 116.2
CAS Number: 124-09-4
MDL Number: MFCD00008243
UNSPSC Code: 12352100
Purity: 0.98
Boiling Point: 204-205 C
Melting Point: 42-45 C (lit.)
Density: 0.89 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.439 (lit.)
Storage: room temp
EINECS Number: 204-679-6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.