Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethylenetetramine, 99% (Methenamine), Reagents

Catalog No. 01337220
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-220 1 kg
01-337-221 100 g
01-337-219 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-220 Supplier Reagents Holdings Llc Supplier No. C22136001KG
Add to Cart
Add to Cart

Hexamethylenetetramine, 99% (Methenamine), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 100-97-0
Molecular Formula C6H12N4
Molecular Weight (g/mol) 140.19
InChI Key VKYKSIONXSXAKP-UHFFFAOYSA-N
Synonym Methenamine, Urotropine, Hexamine
IUPAC Name 1,3,5,7-tetraazatricyclo[3.3.1.1³,⁷]decane
SMILES C1N2CN3CN1CN(C2)C3

Specifications

Boiling Range Sublimes
Melting Point 280°C
Color Colorless/White
Boiling Point Sublimes
Melting Range 280°C
Packaging Poly Bottle
Quantity 1 kg
Specific Gravity 1.331
Grade Reagent
Physical Form Solid
Chemical Name or Material Hexamethylenetetramine, 99% (Methenamine)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.