Learn More
Hexamethylenetetramine, ACS, 99+%
Description
Hexamethylenetetramine is used in the manufacturing of phenolic resins liquid preparations and molding compounds. It finds applications in in brake and clutch linings, abrasive products, non-woven textiles and fireproof materials. It is also used as a drug taken by mouth to prevent recurrent urinary tract infections. It is used as a solid fuel in association with 1,3,5-trioxane in campers, hobbyists, the military and relief organizations. It acts as a food additive and as a preservative. It is an important reagent in organic synthesis, especially Duff reaction, Sommelet reaction and in the Delepine reaction. Further, it is used in adhesives, coatings, sealants, and leather preservatives. It is utilized as a formaldehyde releaser for photographic film development and as a hardener for epoxy resins of the bisphenol A type.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Melting Point | 280°C (sublimation) |
| Color | White |
| Density | 1.33 g/mL |
| Flash Point | 250°C (482°F) |
| Quantity | 500 g |
| UN Number | UN1328 |
| Beilstein | 2018 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,5966 |
| Solubility Information | Soluble in water,dimethyl sulfoxide,chloroform,ethanol and acetone. Slightly soluble in ether. |
| Show More |
Safety and Handling
| Hazard Category | H228-H317-H500 |
| Hazard Statement | GHS H Statement H228-H317 Flammable solid. May cause an allergic skin reaction. |
| Precautionary Statement | P210-P240-P241-P261-P272-P280-P302+P352-P333+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: HEXAMETHYLENETETRAMINE |
| EINECSNumber | 202-905-8 |
| RTECSNumber | MN4725000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.