Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethylphosphoramide, 98%, Thermo Scientific™

Catalog No. AC411520050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC411520050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC411520050 Supplier Thermo Scientific Chemicals Supplier No. 411520050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 680-31-9
Molecular Formula C6H18N3OP
Molecular Weight (g/mol) 179.204
InChI Key GNOIPBMMFNIUFM-UHFFFAOYSA-N
Synonym hexamethylphosphoramide, hmpa, hexametapol, hexamethylphosphoric triamide, hempa, hmpta, hexamethylphosphoric acid triamide, hexamethylphosphorictriamide, hexamethylphosphotriamide, phosphoric hexamethyltriamide
PubChem CID 12679
ChEBI CHEBI:24565
IUPAC Name N-[bis(dimethylamino)phosphoryl]-N-methylmethanamine
SMILES CN(C)P(=O)(N(C)C)N(C)C

Specifications

Alpha Vector Hexamethylph
Color Colorless
Density 1.03g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.4570 to 1.4590 (20°C, 589nm)
Linear Formula OP(N(CH3)2)3
Quantity 5 g
Fieser 01,430; 02,208; 03,149; 04,244; 05,323; 06,273; 07,168; 08,240; 09,235; 10,196; 11,253; 12,239; 13,142; 14,0119; 15,165
Merck Index 15, 4763
Formula Weight 179.2
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Hexamethylphosphoramide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.