accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 680-31-9

IUPAC Name: [bis(dimethylamino)phosphoryl]dimethylamine
Synonyms: hexamethylphosphoramide hexamethylphosphorsaeuretriamid hexamethylphosphoric-triamide hmpta hempa phosphoric hexamethyltriamide hexamethylphosphoramid HMPA HMPT phosphoric tris(dimethylamide)
Molecular Weight (g/mol): 179.2
Molecular Formula: C6H18N3OP
InChi Key: GNOIPBMMFNIUFM-UHFFFAOYSA-N
SMILES: CN(C)P(=O)(N(C)C)N(C)C
Molecular Weight (g/mol) 
  • (7)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (7)
  • (3)
Percent Purity 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (7)
  • (1)
  • (3)
  • (1)
  • (7)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (7)
  • (3)
  • (1)
  • (3)
  • (4)
  • (1)