Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexamethylphosphoramide, 99%, MP Biomedicals

Catalog No. ICN15125180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15125180 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15125180 Supplier MP Biomedicals, Inc Supplier No. 0215125180
Add to Cart
Add to Cart

Chemical Identifiers

CAS 680-31-9
Molecular Formula C6H18N3OP
Molecular Weight (g/mol) 179.204
InChI Key GNOIPBMMFNIUFM-UHFFFAOYSA-N
Synonym hexamethylphosphoramide, hmpa, hexametapol, hexamethylphosphoric triamide, hempa, hmpta, hexamethylphosphoric acid triamide, hexamethylphosphorictriamide, hexamethylphosphotriamide, phosphoric hexamethyltriamide
PubChem CID 12679
ChEBI CHEBI:24565
IUPAC Name N-[bis(dimethylamino)phosphoryl]-N-methylmethanamine
SMILES CN(C)P(=O)(N(C)C)N(C)C

Specifications

Quantity 100 mL
Formula Weight 179.2
Chemical Name or Material Hexamethylphosphoramide, 99%
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.