Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexanamide 98.0+%, TCI America™
SDP

Catalog No. C002425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C002425G 25 g
C00245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C002425G Supplier TCI America Supplier No. C002425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 628-02-4
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00008046
InChI Key ALBYIUDWACNRRB-UHFFFAOYSA-N
Synonym capronamide, hexamide, hexylamide, n-caproamide, caproamide, n-hexanamide, hexamide van, hexanoamide, n-capronamide, n-caproic amide
PubChem CID 12332
IUPAC Name hexanamide
SMILES CCCCCC(=O)N

Specifications

Melting Point 100°C
Color White
Quantity 25 g
Formula Weight 115.18
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Hexanamide
EINECSNumber (2)-0824
RTECSNumber MN7875000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.